LanthaScreen® Tb-anti-AKT [pSer473] Antibody Kit

Catalog number: PV5268

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

LanthaScreen® Tb-labeled phospho-specific antibodies, allow for the detection of intracellular post-translational modifications of specific target proteins. To facilitate in-house kinase assay configuration and screening, we have developed a selection of Tb-labeled phospho-specific antibodies, which may be accessed in small scale and screening quantities.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Clonality: Monoclonal
Concentration: 0.5 mg⁄ml
For Use With (Equipment): Microplate Reader
Form: Liquid
High Throughput Compatibility: High Throughput-Compatible
Host Species: Rabbit
Label or Dye: Tb (Terbium)
Product Line: LanthaScreen®
Product Size: 1 mg
Purification: Purified
Reactivity: Human
SubClass (Isotype): IgG
Shipping Condition: Dry Ice

Contents & storage

Terbium labeled anti-AKT [pSer473] antibody in HEPES buffered saline (137 mM NaCl, 2.7 mM KCl, 10 mM HEPES pH 7.5). Storage is at -20°C

Documents

Manuals & protocols