Thermo Scientific™

LanthaScreen™ Tb-anti-ERK2 [pThr185/pTyr187] Antibody Kit

Catalog number: PV5270
Thermo Scientific™

LanthaScreen™ Tb-anti-ERK2 [pThr185/pTyr187] Antibody Kit

Catalog number: PV5270
Catalog Number
PV5270
Unit Size
1 mg
Price (USD)
18,630.00
Your Price:
Availability
***
Quantity
Catalog Number
Unit Size
25 µg
Price (USD)
3,400.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV52701 mg
18,630.00
Your Price:
***
PV526925 µg
3,400.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
To facilitate in-house kinase assay configuration and screening, we have developed a selection of Tb-labeled phospho-specific antibodies, which may be accessed in small scale and screening quantities.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
0.5 mg/mL, 0.5 mg⁄ml, 0.5 mg/ml
Host Species
Rabbit
Purification Method
Purified
Quantity
1 mg
Antibody Conjugate
Lanthanide Chelates, Terbium (Tb)
Antibody Fragment
Whole Antibody
Clonality
Monoclonal
For Use With (Equipment)
Microplate Reader
Form
Liquid
High-throughput Compatibility
High-throughput Compatible
Host abrv
Rb
Label or Dye
Tb (Terbium)
Label Type
Lanthanide Chelates
Marker
0.5 mg⁄ml
Reactivity
Human
Shipping Condition
Dry Ice
Reactivity abrv
Hu
Sub Class Isotype
IgG
Product Line
LanthaScreen™

Contents & Storage

Terbium labeled anti-ERK2 [pThr185⁄pTyr187] antibody in HEPES buffered saline (137 mM NaCl, 2.7 mM KCl, 10 mM HEPES pH 7.5). Storage is at -20 °C

Figures

Frequently asked questions (FAQs)

Citations & References