LanthaScreen™ Tb-anti-PRAS40 [pSer183] Antibody

Catalog number: PV5579

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

A part of the PI3K⁄AKT Pathway, PRAS40 (Proline-rich AKT substrate, 40 kDa) is a cytosolic protein that is phosphorylated at position Ser183. PRAS40 has been shown to bind to the mTORC1 complex and inhibit downstream signaling (i.e., phosphorylation of 4E-BP1 or p70S6K).
For use in combination with the LanthaScreen® PRAS40 HEK293E cell line (K1528) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Excitation⁄Emission (nm): 340⁄490, 520
Label or Dye: Tb (Terbium)
Product Line: LanthaScreen®
Product Size: 1 mg
Reactivity: Human
Readout: End Point
Target Gene: Ser/Thr Kinases
Shipping Condition: Dry Ice

Contents & storage

Terbium-labeled pPRAS40 (pSer183) antibody in HEPES buffered saline. Store at -20°C.

Documents

Manuals & protocols