Product Image
Thermo Scientific™

Kinase Tracer 178, 25 μM

In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or “tracer" such as Kinase Tracer 178Read more
Have Questions?
Catalog NumberQuantity
PV559325 μL
Catalog number PV5593
Price (USD)
670.00
Each
Quantity:
25 μL
Price (USD)
670.00
Each
In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or “tracer" such as Kinase Tracer 178 to a kinase is detected by addition of a Eu-labeled anti-tag antibody. Binding of the tracer & antibody to a kinase results in a high degree of FRET, whereas displacement of the tracer with a kinase inhibitor results in a loss of FRET. Unlike many kinase activity assays, this assay is a simple mix and read assay, with no development steps.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayKinase Binding Assay
Assay EntryBiochemical binding/kinetic
For Use With (Application)TR-FRET
For Use With (Equipment)Microplate Reader
Format384-well plate
Label or DyeAlexa Fluor 647
Product TypeKinase Tracer
Quantity25 μL
ReadoutKinetic
Shipping ConditionDry Ice
TargetKinases (Multiple)
Target EntryPTK6, DDR2, BMPR1A, RAF1 (cRAF) Y340D Y341D, ACVR1B, WNK2, TEC, DDR1, EPHB4, ERBB4, KIT V654A, TGFBR1, NLK, TGFBR2, EPHB3, BRAF V599E, BRAF, ACVR2B, ACVR2A
Product LineLanthaScreen
Unit SizeEach
Contents & Storage
Store at -20°C.