BacMam Enhancer Solution (1000X) - FAQs
BacMam Enhancer Solution (1000X) - FAQs
View additional product information for BacMam Enhancer Solution (1000X) - FAQs (PV5835)
1 product FAQs found
I am using your CellLight BacMam reagents for fluorescent protein-labeling of my cell organelles, but they aren't labeling well. How can I improve it?
First, make sure that your cells are mammalian and non-blood derived. If the cells have low transduction efficiency (i.e., only a few cells are labeling or are labeling only dimly), first try increasing the particle number per cell. The next thing to try is to use the BacMam Enhancer Solution. If, however, your cells show labeling, but the localization seems to be too generalized or ubiquitous within the cell, then you should reduce the particle number per cell. The manual gives a guideline and formula for calculating particle number per cell.