Biotin-Anti-His-Tag Antibody
Invitrogen™

Biotin-Anti-His-Tag Antibody

Biotin-Anti-His-Tag Antibody
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PV60901 mg
PV608925 μg
Catalog number PV6090
Price (USD)
15,726.65
Online Exclusive
16,910.00
Save 1,183.35 (7%)
Each
Quantity:
1 mg
Price (USD)
15,726.65
Online Exclusive
16,910.00
Save 1,183.35 (7%)
Each
Biotin-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using biotin negatively impacts protein activity. These antibodies can be used with our Kinase Tracers and Eu-Streptavidin to conduct LanthaScreen™ Eu Kinase Binding Assays. Learn More and See Which Kinases Have Been Validated Using This Assay
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.5 mg⁄ml
DescriptionBiotin-Anti-His-Tag Antibody, 1mg, 0.5mg/mL Concentration, Biotin Conjugate, HEPES buffered saline (137mM NaCl, 2.7mM KCl, 10mM HEPES pH 7.5) Storage Buffer, Used for Pharma and Biopharma, Target and Lead Identification and Validation
Molecular Weight of Antigen150kDa
Packaging1 Each
Primary or SecondaryPrimary
Research DisciplineKinase Biology
Shipping ConditionDry Ice
ApplicationsKinase Assay, Kinase Assay
ConjugateBiotin
FormFrozen
Quantity1 mg
Regulatory StatusRUO, RUO
Unit SizeEach
Contents & Storage
Biotin-labeled anti-His tag antibody in HEPES buffered saline. Store at -20°C.