Qdot™ 655 ITK™ Streptavidin Conjugate Kit
Invitrogen™

Qdot™ 655 ITK™ Streptavidin Conjugate Kit

Das Qdot™ 655 ITK™ Streptavidin-Konjugat-Kit enthält das Qdot™ Streptavidin-Konjugat als 250 µl einer 2 µM-Lösung. Anders als die Qdot™ Streptavidin-KonjugateWeitere Informationen
Have Questions?
KatalognummerMenge
Q10021MP1 Kit
Katalognummer Q10021MP
Preis (EUR)
990,00
Each
Menge:
1 Kit
Preis (EUR)
990,00
Each
Das Qdot™ 655 ITK™ Streptavidin-Konjugat-Kit enthält das Qdot™ Streptavidin-Konjugat als 250 µl einer 2 µM-Lösung. Anders als die Qdot™ Streptavidin-Konjugate haben die Qdot™ ITK™ Streptavidin-Konjugate den Streptavidin-Kovalent ohne einen PEG-Linker an der inneren amphiphilen Beschichtung angebracht. Diese Qdot™ Konjugate können für die Bindung biotinylierter Sonden verwendet werden. Das Kit enthält außerdem 30 ml Qdot™ Inkubationspuffer.
Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.
Specifications
Konzentration2 μM
ProdukttypStreptavidin-Konjugat (fluoreszierend)
Menge1 Kit
VersandbedingungRaumtemperatur
KonjugatQdot 655
ProduktlinieITK, Qdot
Unit SizeEach
Inhalt und Lagerung
Im Kühlschrank lagern (2 bis 8 °C).

Häufig gestellte Fragen (FAQ)

I am getting very high background with my Qdot streptavidin conjugate. Do you have any suggestions?

Here are some suggestions: Use the Qdot Incubation Buffer (Cat. No. Q20001MP). The included buffer is formulated specifically for improved signal-to-background ratios in most immunolabeling applications using the Qdot streptavidin conjugates. Alternate buffers may result in more variable staining and, in particular, may increase background staining. However, some specific applications may require other buffer conditions. Please see the protocol "Double-labeling Using Qdot Streptavidin conjugates."
Determine if the sample has a high level of endogenous biotin. Block the sample using an avidin-biotin pre-blocking step.
If you have used the Qdot Incubation Buffer and still get high nonspecific background, then it may be necessary to check other steps of your procedure. Blocking the sample with BSA or normal animal serum will generally decrease nonspecific binding of both antibodies and Qdot streptavidin conjugates. It is a good practice to dilute your primary and secondary antibodies in the blocking buffer. Some tissues such as spleen and kidney sections may contain endogenous biotin, which may contribute to non-specific signal. Endogenous biotin can be blocked with an avidin/biotin blocking kit (Cat. No. E21390).
Grainy staining or clumps of fluorescent material appear in the background.
Occasionally the BSA within the Qdot Incubation Buffer shows slight aggregation over time. It is necessary to remove this aggregate prior to labeling the sample with the Qdot streptavidin conjugate. Spin down the incubation mixture before addition to the sample. This can be accomplished by spinning the samples in a benchtop centrifuge (Eppendorf 5415) at 5,000 x g for 2 minutes. The material can also be passed over a 0.2 µm spin filter unit before you add it to the sample for staining to remove microscopic precipitates. If you are using a buffer that is different than the Qdot Incubation Buffer, this behavior can often be attributed to higher levels of NaCl or other salts in the incubation buffer, and may not be easily fixed with filtration. In this case, reduce the overall salt concentration.
Optimize concentration of biotinylated secondary antibodies.
Optimizing specific signal can often be achieved by adjusting the level of biotinylated antibody used instaining. High levels of biotinylated antibody are necessary to obtain specific labeling, but overly high levels will contribute to nonspecific binding of the antibody to the sample. Nonspecifically bound biotinylated antibody will bind to the Qdot streptavidin conjugate, resulting in higher staining of the background.
Optimize concentration of Qdot streptavidin conjugate.
Just as titration of primary and secondary antibodies is necessary to achieve optimal specific signal in immunolabeling applications, the level of the final probe should be optimized for each conjugate. In general, concentrations at or slightly below saturation should have the optimal signal-to-background ratio, while concentrations substantially higher than saturation will compromise the assay with higher background levels.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am getting no signal with my Qdot streptavidin conjugate. What should I do?

Here are some suggestions:

Confirm imaging/detection setup suitability.
Make sure that you are using an appropriate filter set to detect the signal. Please consult Table 1 in the Qdot Biotin User Manual for a list of appropriate and optimal filters.
Check to see that Qdot conjugate is fluorescing using an alternative light source.
Qdot conjugates will normally fluoresce brightly under a hand-held ultraviolet lamp (long wave, such as the type used to visualize ethidium bromide on agarose gels). Although we have not seen pronounced loss of fluorescence of these materials under any storage conditions that we have investigated, we have not been able to examine all storage conditions. If the Qdot product does not appear to fluoresce under the long wave UV excitation, please contact Technical Support at techsupport@qdots.com. For a microscope, perform a spot test: place a small droplet (2 to 5 µL) of the quantum dot solution onto a clean slide (no coverslip) and examine under the appropriate filter set at low magnification.
Confirm the specificity and titer of primary antibody.
Make sure the antibody will recognize the intended targets. Make sure there is sufficient primary antibody bound to the targets. This verification can be performed by ELISA-based capture of the antigen of interest, or by other techniques that can be found in lab manuals such as the Current Protocols in Immunology.
For Qdot streptavidin conjugates, confirm biotinylation of antibody.
Make sure your antibodies are effectively biotinylated. It may be necessary to independently adjust the concentration of both the primary and secondary antibodies used in the assay to obtain optimal signal and minimal background.
PAP pen ink may quench signal.
Use an alternate method for isolating target areas on the slide. If your protocol requires the use of a PAP pen, we recommend the ImmEdge Hydrophobic Barrier Pen (Cat. No. H-4000) from Vector Labs.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the best way to remove white precipitate from my ITK Qdot nanocrystals?

Spinning your ITK Qdot nanocrystals at approximately 3,000 rpm for 3-5 minutes should remove the white precipitate from the supernatant. Use the supernatant immediately.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I see a white precipitate in my ITK Qdot nanocrystals; should I be concerned?

The precipitate in the organic ITK Qdot nanocrystals occurs with some frequency. The ITK Qdot nanocrystals sometimes include impurities that show as a white precipitate.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Why do my Qdot nanocrystals appear to be blinking?

Blinking is an inherent property of quantum dots; in fact, all single-luminescent molecules blink, including organic dyes. The brightness and photostability of Qdot nanocrystals makes the blinking more visibly apparent. Under higher energy excitation, Qdot nanocrystals blink even faster.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Zitierungen und Referenzen (5)

Zitierungen und Referenzen
Abstract
Correlated light and electron microscopic imaging of multiple endogenous proteins using Quantum dots.
Authors:Giepmans BN, Deerinck TJ, Smarr BL, Jones YZ, Ellisman MH
Journal:Nat Methods
PubMed ID:16179920
'The importance of locating proteins in their context within cells has been heightened recently by the accomplishments in molecular structure and systems biology. Although light microscopy (LM) has been extensively used for mapping protein localization, many studies require the additional resolution of the electron microscope. Here we report the application ... More
Multiplexed hybridization detection of quantum dot-conjugated DNA sequences using surface plasmon enhanced fluorescence microscopy and spectrometry.
Authors:Robelek R, Niu L, Schmid EL, Knoll W
Journal:Anal Chem
PubMed ID:15481968
'In this study, the general suitability of quantum dot (QD)-DNA conjugates for the surface plasmon enhanced fluorescence spectroscopy technique is demonstrated. Furthermore, the QD-DNA system is transferred to the platform of surface plasmon enhanced fluorescence microscopy. Using this technique together with a microarray format, in which the sensor-bound single-stranded catcher ... More
Near-complete suppression of quantum dot blinking in ambient conditions.
Authors:Hohng S, Ha T
Journal:J Am Chem Soc
PubMed ID:14759174
Continuous distribution of emission states from single CdSe/ZnS quantum dots.
Authors:Zhang K, Chang H, Fu A, Alivisatos AP, Yang H,
Journal:Nano Lett
PubMed ID:16608295
The photoluminescence dynamics of colloidal CdSe/ZnS/streptavidin quantum dots were studied using time-resolved single-molecule spectroscopy. Statistical tests of the photon-counting data suggested that the simple
Blinking and nonradiant dark fraction of water-soluble quantum dots in aqueous solution.
Authors:Yao J, Larson DR, Vishwasrao HD, Zipfel WR, Webb WW
Journal:Proc Natl Acad Sci U S A
PubMed ID:16169907
Water-soluble quantum dots (qdots) are now being used in life sciences research to take advantage of their bright, easily excited fluorescence and high photostability. Although the frequent erratic blinking and substantial dark (never radiant) fractions that occur in all available qdots may interfere with many applications, these properties of individual ... More