Qdot™ 655 Biotin Conjugate Kit
Invitrogen™

Qdot™ 655 Biotin Conjugate Kit

The biotin-labeled Qdot™ 655 nanocrystals are available for detecting streptavidin probes or for creating noncovalent conjugates with streptavidin-labeled molecules orRead more
Have Questions?
Catalog NumberQuantity
Q10321MP1 kit
Catalog number Q10321MP
Price (USD)
707.65
Online exclusive
944.00
Save 236.35
Each
Quantity:
1 kit
Price (USD)
707.65
Online exclusive
944.00
Save 236.35
Each
The biotin-labeled Qdot™ 655 nanocrystals are available for detecting streptavidin probes or for creating noncovalent conjugates with streptavidin-labeled molecules or with other biotinylated molecules using a streptavidin bridge. The product is provided as 250 μL of a 2 μM solution and includes 30 mL of Qdot™ incubation buffer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeBiotin Qdot Conjugation Kit
Quantity1 kit
Shipping ConditionRoom Temperature
Product LineQdot
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

What is the best way to remove white precipitate from my ITK Qdot nanocrystals?

Spinning your ITK Qdot nanocrystals at approximately 3,000 rpm for 3-5 minutes should remove the white precipitate from the supernatant. Use the supernatant immediately.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I see a white precipitate in my ITK Qdot nanocrystals; should I be concerned?

The precipitate in the organic ITK Qdot nanocrystals occurs with some frequency. The ITK Qdot nanocrystals sometimes include impurities that show as a white precipitate.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Why do my Qdot nanocrystals appear to be blinking?

Blinking is an inherent property of quantum dots; in fact, all single-luminescent molecules blink, including organic dyes. The brightness and photostability of Qdot nanocrystals makes the blinking more visibly apparent. Under higher energy excitation, Qdot nanocrystals blink even faster.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

My Qdot nanocrystals were brightly fluorescent before I mounted my samples; now I'm seeing a loss of fluorescence. Why is this happening?

Appropriate mounting media selection is very important to retain the fluorescence of Qdot nanocrystals. In our studies, Qdot nanocrystals work best with the following mountants:

HistoMount medium (Cat No. 00-8030); best for long term archiving
Cytoseal 60 Mountant
Clarion Mountant
Most polyvinyl alcohol-based mountants (limited storage time, less than weeks)
Water-based mountants (limited storage time, less than week)
Up to 50% glycerol (limited storage time, less than week)
Note: We do not recommend using ProLong mounting media with Qdot nanocrystals as it will quench their fluorescence.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Why can't I freeze my Qdot nanocrystal solution?

Freezing will cause the product to aggregate. The Qdot nanocrystals cannot be dispersed into solution after aggregation.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (4)

Citations & References
Abstract
Separation of bioconjugated quantum dots using capillary electrophoresis.
Authors:Vicente G, Colón LA,
Journal:Anal Chem
PubMed ID:18278946
'Capillary electrophoresis (CE) with laser-induced fluorescence (LIF) detection was used to separate different bioconjugated CdSe/ZnS quantum dots (QDs). The QD nanocrystals studied were conjugated to the biomolecules streptavidin, biotin, and immunoglobulin G. The bioconjugated QDs showed different electrophoretic mobilities, which appear to depend upon the biomolecule that is attached to ... More
Quantum dots as efficient energy acceptors in a time-resolved fluoroimmunoassay.
Authors:Hildebrandt N, Charbonnière LJ, Beck M, Ziessel RF, Löhmannsröben HG
Journal:Angew Chem Int Ed Engl
PubMed ID:16247812
Lanthanides to quantum dots resonance energy transfer in time-resolved fluoro-immunoassays and luminescence microscopy.
Authors:Charbonnière LJ, Hildebrandt N, Ziessel RF, Löhmannsröben HG,
Journal:J Am Chem Soc
PubMed ID:17002375
A time-resolved fluoro-immunoassay (TR-FIA) format is presented based on resonance energy transfer from visible emitting lanthanide complexes of europium and terbium, as energy donors, to semiconductor CdSe/ZnS core/shell nanocrystals (quantum dots, QD), as energy acceptors. The spatial proximity of the donor-acceptor pairs is obtained through the biological recognition process of ... More
Single particle quantum dot imaging achieves ultrasensitive detection capabilities for Western immunoblot analysis.
Authors:Scholl B, Liu HY, Long BR, McCarty OJ, O'Hare T, Druker BJ, Vu TQ,
Journal:ACS Nano
PubMed ID:19453141
Substantially improved detection methods are needed to detect fractionated protein samples present at trace concentrations in complex, heterogeneous tissue and biofluid samples. Here we describe a modification of traditional Western immunoblotting using a technique to count quantum-dot-tagged proteins on optically transparent PVDF membranes. Counts of quantum-dot-tagged proteins on immunoblots achieved ... More