QUBIT QUANTITATION LAB STARTER - FAQs
QUBIT QUANTITATION LAB STARTER - FAQs
View additional product information for QUBIT QUANTITATION LAB STARTER - FAQs (Q32863)
1 product FAQs found
Do you offer standard #2 that is included in the Qubit dsDNA BR Assay Kit as a stand-alone product?
Standard #2 of the Qubit dsDNA BR Assay Kit can be obtained separately as Qubit 1X dsDNA BR Assay Lambda Standards (Cat No. Q33263). Standard #1 is also included.