Qubit® 2.0 Fluorometer International Power Cord - FAQs
Qubit® 2.0 Fluorometer International Power Cord - FAQs
View additional product information for Qubit® 2.0 Fluorometer International Power Cord - FAQs (Q32868)
1 product FAQs found
How does the accuracy and sensitivity of the Qubit quantitation assays using the Qubit fluorometer compare to a microplate reader?
The accuracy and sensitivity of the Qubit quantitation assays are the same as that of a microplate reader. This was a requirement during product development. The detection limits for each Qubit kit can be found on the corresponding product manual, which can be found by searching our website by keyword or catalog number.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.