Qubit™ Endotoxin Detection Assay Kit - FAQs
Qubit™ Endotoxin Detection Assay Kit - FAQs
View additional product information for Qubit™ Endotoxin Detection Assay Kit - FAQs (Q32891)
3 product FAQs found
Can I use the Qubit Endotoxin Detection Assay Kit on the Qubit 4 Fluorometer?
No, the Qubit Endotoxin Detection Assay Kit is designed for use with the Qubit Flex Fluorometer and is not currently for use on the Qubit 4 Fluorometer.
To use the Qubit Endotoxin Detection Assay Kit on the Qubit Flex Fluorometer, do I need to perform a software update ?
Yes, the endotoxin detection assay program will be available on Qubit Flex Software version 1.8.0 or later. The latest software is available under the Resources section of www.thermofisher.com/qubit
How do I store the Qubit Endotoxin Detection Assay Kit?
The entire kit can be stored at 2-8 degrees C for 6 months. Reconstituted Endotoxin standard solutions are stable for 4 weeks at 2-8 degrees C and reconstituted dye can be stored for 4 weeks at 2-8 degrees C. Reconstituted amebocyte lysate solution is stable for 1 week at -20 degrees C or colder if frozen immediately after reconstitution.