Qubit™ 1X dsDNA HS Assay Kit, 100 assays - FAQs
Qubit™ 1X dsDNA HS Assay Kit, 100 assays - FAQs
View additional product information for Qubit™ 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits - FAQs (Q33231, Q33230, Q33266, Q33265)
3 product FAQs found
What is the expiration date for Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits (Cat. Nos. Cat. Q33230, Q33231, Q33265, Q33266)?
The Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits (Cat. Nos. Cat. Q33230, Q33231, Q33265, Q33266) are stable for 6 months from the date of receipt when stored as directed.
For more information on product storage, please refer to the user guides:
Qubit 1X dsDNA HS Assay Kit and Qubit 1X dsDNA BR Assay Kit
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
What is the difference between The Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits (Cat. Nos. Q33230, Q33231, Q33265, Q33366) and Qubit dsDNA Quantification Assay Kits (Cat. Nos. Q32850, Q32851, Q32853, Q32854)?
The Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits (Cat. Nos. Q33230, Q33231, Q33265, Q33366) are newer and have a simplified workflow compared to the original Qubit dsDNA Quantification Assay Kits (Cat. Nos. Q32850, Q32851, Q32853, Q32854). The original Qubit kits contain separate dyes and buffer components that must be mixed together to make a working solution, in which the dye degrades after a few hours. With the newer Qubit 1X dsDNA Assay kits, the dye is premixed with a buffer that keeps the dye stable long-term and can be added directly to DNA samples.
You can find more information about the Qubit 1X dsDNA assay, by clicking on the Technical Note provided below:
Qubit 1X dsDNA assays: simplified workflow and improved performance
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
How should I store my Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits (Cat. No. Q33230, Q33231, Q33265, Q33266)?
The Qubit 1X dsDNA High Sensitivity (HS) (Cat. Nos. Q33230, Q33231) and Broad Range (BR) Assay Kits (Cat. Nos. Q33265, Q33266) have different storage conditions.
For Qubit 1X dsDNA HS Assay Kits (Cat. Nos. Q33230, Q33231), store all components at 2 - 8 degrees C.
For Qubit 1X dsDNA BR Assay Kits (Cat. Nos. Q33265, Q33266), store the working solution at 18 - 28 degrees C. Store the standards at 2 - 8 degrees C.
The assay kits are stable for at least 6 months from the date of receipt.
For more information, please refer to these user guides: Qubit 1X dsDNA HS Assay Kit and Qubit 1X dsDNA BR Assay Kit
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.