Product Image
Invitrogen™

Qubit™ 4 System Verification Assay Kit

The Qubit 4 System Verification Assay Kit is a fast, easy-to-use, reagent-based assay that tests the performance of the QubitRead more
Have Questions?
Catalog NumberQuantity
Q3323750 assays
Catalog number Q33237
Price (USD)
165,36
Each
Quantity:
50 assays
Price (USD)
165,36
Each
The Qubit 4 System Verification Assay Kit is a fast, easy-to-use, reagent-based assay that tests the performance of the Qubit 4 Fluorometer. The kit consists of three components: a blank reagent solution, a green fluorescent reagent, and a far-red fluorescent reagent. The green fluorescent reagent is designed to test the blue excitation (470 nm LED; 430-495 nm excitation filter) and green fluorescence detection (510-580 nm emission filter) on the fluorometer, while the far-red fluorescent reagent is designed to test the red excitation (635 nm LED; 600-645 nm excitation filter) and far-red fluorescence detection (665-720 nm emission filter).

The assay kit provides sufficient reagents to perform up to 50 assays.

Note: The Qubit 4 System Verification Assay Kit can only be used with the Qubit 4 Fluorometer.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineQubit
Quantity50 assays
Shipping ConditionRoom Temperature
For Use With (Equipment)Qubit 4 Fluorometer
TypeSystem Verification Assay Kit
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

What routine maintenance is recommended for the Qubit 4 Fluorometer, with WIFI (Cat. No. Q33238)?

The Qubit 4 Fluorometer, with WIFI (Cat. No. Q33238), does not require any specific routine maintenance. However, based on customer experience, we recommend the following:

  • From time to time, it may be useful to check the opening of the Qubit for dust particles and dirt. If needed, you can use a lint-free cloth or lenses cleaning paper towels to wipe the inside of the Qubit with some 75%-95% ethanol.

  • If you experience inaccurate measurements, regardless of the kit you use, we recommend testing the light source of the instrument with the Qubit 4 System Verification Assay Kit (Cat. No. Q33238).


Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.

How should I store my Qubit 4 Assay Kit for System Verification (Cat. No. Q33237)?

Qubit 4 Assay Kit for System Verification (Cat. No. Q33237) should be stored at room temperature and is stable for 12 months from receipt.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center .

Can I use the Qubit 4 Fluorometer System Verification Assay Kit on previous models of the Qubit fluorometer?

Unfortunately, this is not possible. The Qubit 4 Fluorometer System Verification Assay Kit was designed for use only with the Qubit 4.0 fluorometer.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the buffer used for Components A, B, and C within the Qubit 4 Fluorometer System Verification Assay Kit?

The composition/formulation of solutions in Components A, B and C within the Qubit 4 Fluorometer System Verification Assay Kit is proprietary.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I accidentally refrigerated/froze the Qubit 4 Fluorometer System Verification Assay Kit. Can I still use the kit?

The components in this kit may still be usable if accidentally refrigerated or frozen, but we would recommend examining the solutions for any turbidity or precipitation. If cloudiness or if any precipitates are visible, this product should not be used.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.