Qubit™ 4 Starter Packages, with WiFi
Invitrogen™

Qubit™ 4 Starter Packages, with WiFi

Accurately quantify protein, dsDNA, and RNA with the Qubit 4 Protein BR, Quantitation, NGS, and RNA IQ Starter Kits, which feature a fluorescent dye that emits signal only when bound to its target molecule, minimizing the effect of contaminants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
Q33241RNA IQ Starter Kit, with WiFi
Q33239Quant Starter Kit, w/ WiFi
Q33240NGS Starter Kit, w/ WiFi
A51292Prot. Broad Range Starter Kit
Catalog number Q33241
Price (USD)
3,789.50
Special offer
Online exclusive
Ends: 04-Dec-2026
5,830.00
Save 2,040.50
Each
Type:
RNA IQ Starter Kit, with WiFi
Price (USD)
3,789.50
Special offer
Online exclusive
Ends: 04-Dec-2026
5,830.00
Save 2,040.50
Each
Achieve accurate and reliable results with Qubit 4 Starter Packages, which combine the Qubit 4 Fluorometer with different assay kits for the quantification of nucleic acids or proteins. Qubit reagents bind selectively to DNA, RNA, or protein and only emit a fluorescent signal when bound to the target. This minimizes the effects of contaminants on the result and ensures accurate readings for sensitive samples.

With Qubit assays, the concentration or quality of the target molecule in the sample is reported by a fluorescent dye that emits a signal only when bound to its target, minimizing the effects of contaminants on the result. Whether you are an expert or a novice, the easy-to-use touchscreen menus make it easy to perform assays, with accurate and reliable results displayed in just a few seconds. Qubit 4 with Wi-Fi also provides flexible data exportation using a USB drive, cloud connectivity, or direct USB cable connection so quantification data can be easily accessed.

Qubit 4 Quantitation Starter Kit

The Qubit 4 With Qubit assays, the concentration or quality of the target molecule in the sample is reported by a fluorescent dye that emits a signal only when bound to its target, minimizing the effects of contaminants on the result. Whether you are an expert or a novice, the easy-to-use touchscreen menus make it easy to perform assays, with accurate and reliable results displayed in just a few seconds. Qubit 4 with Wi-Fi also provides flexible data exportation using a USB drive, cloud connectivity, or direct USB cable connection so quantification data can be easily accessed.

Qubit 4 Quantitation Starter Kit (Cat. No. Q33239) includes:

  • Qubit 4 Flurometer (Cat. No. Q33226)
  • Qubit Assay Tubes, Set of 500 (Cat. No. Q32856)
  • Qubit 1X dsDNA HS, 100 Assays (Cat. No. Q33230)
  • Qubit RNA Assay Kit, 100 (Cat. No. Q32852)
  • Qubit RNA IQ Kit, 75 Assays (Cat. No. Q33221)
  • Qubit RNA BR Assay Kit, 100 (Cat. No. Q10210)
  • Qubit 4 System Verification Assay Kit (Cat. No. Q33237)
  • Kit, 802.11AC/B/G/N Wi-Fi Dongle (Cat. No. A26774)
  • Qubit 1X dsDNA BR Working Sol (Cat. No. Q33260)
  • Qubit 1X dsDNA BR Standards (Cat. No. Q33261)
  • Qubit Protein BR Assay Kit (Cat. No. A50668)

Qubit 4 NGS (Next-Generation Sequencing) Starter Kit

The Qubit NGS Starter Kit (Cat. No. Q33240) includes:

  • Qubit 4 Fluorometer with WiFi (Cat. No. Q33238)
  • Wi-Fi Dongle (Cat. No. A26774)
  • Qubit Assay Tubes, set of 500 (Cat. No. Q32856)
  • Qubit 1X dsDNA HS Assay Kit, 500 assays (Cat. No. Q33231)

Qubit Protein BR Starter Kit

The Qubit Protein BR Assay Kit provides a quick, easy, and reliable method for determining the protein concentration of purified proteins, lysates, serum, plasma, and complex protein mixtures. The assay can deliver accurate protein quantification across a broad range (0.1–20 mg/mL), enabling most samples to be used neat (undiluted) and eliminating the guess work and dilution steps that accompany traditional protein quantitation methods.

The Qubit Protein BR Assay Kit is a fluorescence-based assay intended for use with the Qubit 4 Fluorometer. The assay is not designed for use with other Qubit models. The kit comes with two calibration standards, Protein BR Assay Buffer, and Protein BR Assay Reagent. Simply add 150–160 μL of Protein BR Assay Buffer to 10 or 20 μL of your sample, followed by 30 μL of Protein BR Assay Reagent. Incubate for 10 minutes at room temperature. Then, insert your sample into a Qubit 4 instrument and read the concentration.

The Qubit 4 Protein BR Starter Kit (Cat. No. A51292) includes:

  • Qubit 4 Fluorometer with WiFi (Cat. No. Q33238)
  • Wi-Fi Dongle (Cat. No. A26774)
  • Qubit Assay Tubes, set of 500 (Cat. No. Q32856)
  • Qubit Protein BR Assay Kit, 100 assays (Cat. No. A50668)

Qubit 4 RNA IQ Starter Kit

The Qubit RNA IQ assay provides a fast, simple method to check whether an RNA sample has degraded. The assay utilizes two unique dyes: one that binds to large, intact RNA, and another that selectively binds to small, degraded RNA. Together they enable you to quickly assess the quality and integrity of an RNA sample. To use, simply add your samples to the RNA IQ working solution, then measure on the Qubit 4 Fluorometer.

The Qubit 4 RNA IQ Starter Package (Cat. No. Q33241) includes:

  • Qubit 4 Fluorometer with WiFi (Cat. No. Q33238)
  • Qubit Assay Tubes, set of 500 (Cat. No. Q32856)
  • Qubit RNA IQ Assay Kit, 275 assays (Cat. No. Q33222)
  • Wi-Fi Dongle (Cat. No. A26774)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)sdDNA Quantification, RNA Quantification, Protein Quantification
IncludesFluorometer, Assay Tubes, Wi-Fi Dongle, Assay Kit
Quantity1 kit
Sample Volume1 to 20 μL
Excitation Wavelength470 nm (Blue LED), 635 nm (Red LED)
For Use With (Equipment)Qubit Fluorometer
Product LineQubit
TypeRNA IQ Starter Kit, with WiFi
Unit SizeEach

Frequently asked questions (FAQs)

I would like to use my Qubit 4 Fluorometer with the Qubit 1X dsDNA BR Assay, but the assay is not listed as an option on the fluorometer. How can I resolve this?

The Qubit 4 Fluorometer needs to be updated. The Qubit 4 Fluorometer can be updated with the USB that was provided with the Qubit. If you do not have the USB, we recommend using an empty 2.0GB that is FAT32 formatted. If the USB is not recognized, leave USB plugged in for a few minutes before running the update. This should give you the assay you need to run the Qubit Assay.

You can find the updates on the Technical Resources for Qubit Fluorometers page.

Which is the best DNA or RNA quantification instrument?

All of the DNA instruments offered by Thermo Fisher Scientific are of excellent quality. To find the right instrument to quantify DNA or RNA in your lab, explore the detailed comparison of UV-Vis spectrophotometers and fluorometers. Things like sensitivity, throughput and budget may be initial considerations when selecting a DNA quantification instrument. You may also consider whether you need target specificity, sample purity information, RNA quality information, or broad dynamic range.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.

What is the difference between UV and fluorescence DNA quantification? Which DNA quantification technology should I choose?

UV and fluorescence technologies work differently to quantify DNA. UV quantification relies on the intrinsic absorptivity of DNA and RNA molecules, while fluorescence quantification uses dyes that specifically bind to your molecule of choice. With UV technology, quantification isn't as sensitive, but it has a broader dynamic range and also gives data about sample purity‒plus it is faster because there is no reagent prep. With fluorescence technology you get higher sensitivity and molecule-specific data, but it has lower dynamic range and reagent prep is required. The technology you need will depend on what features are important for your lab and experimentation.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.

Citations & References (16)

Citations & References
Abstract
A comparative analysis of drinking water employing metagenomics.
Authors:Brumfield KD, Hasan NA, Leddy MB, Cotruvo JA, Rashed SM, Colwell RR, Huq A
Journal:PLoS One
PubMed ID:32271799
'The microbiological content of drinking water traditionally is determined by employing culture-dependent methods that are unable to detect all microorganisms, especially those that are not culturable. High-throughput sequencing now makes it possible to determine the microbiome of drinking water. Thus, the natural microbiota of water and water distribution systems can ... More
Single-cell transcriptomics of the naked mole-rat reveals unexpected features of mammalian immunity.
Authors:Hilton HG, Rubinstein ND, Janki P, Ireland AT, Bernstein N, Fong NL, Wright KM, Smith M, Finkle D, Martin-McNulty B, Roy M, Imai DM, Jojic V, Buffenstein R
Journal:PLoS Biol
PubMed ID:31751331
'The immune system comprises a complex network of specialized cells that protects against infection, eliminates cancerous cells, and regulates tissue repair, thus serving a critical role in homeostasis, health span, and life span. The subterranean-dwelling naked mole-rat (NM-R; Heterocephalus glaber) exhibits prolonged life span relative to its body size, is ... More
Distinct transcriptional roles for Histone H3-K56 acetylation during the cell cycle in Yeast.
Authors:Topal S, Vasseur P, Radman-Livaja M, Peterson CL
Journal:Nat Commun
PubMed ID:31558720
'Dynamic disruption and reassembly of promoter-proximal nucleosomes is a conserved hallmark of transcriptionally active chromatin. Histone H3-K56 acetylation (H3K56Ac) enhances these turnover events and promotes nucleosome assembly during S phase. Here we sequence nascent transcripts to investigate the impact of H3K56Ac on transcription throughout the yeast cell cycle. We find ... More
Characterization of exosomes in peritoneal fluid of endometriosis patients.
Authors:Nazri HM, Imran M, Fischer R, Heilig R, Manek S, Dragovic RA, Kessler BM, Zondervan KT, Tapmeier TT, Becker CM
Journal:Fertil Steril
PubMed ID:32106990
'To demonstrate the feasibility of studying exosomes directly from peritoneal fluid, we isolated exosomes from endometriosis patient samples and from controls, and characterized their cargo.'
Seasonal Succession of Free-Living Bacterial Communities in Coastal Waters of the Western Antarctic Peninsula.
Authors:Luria CM, Amaral-Zettler LA, Ducklow HW, Rich JJ
Journal:Front Microbiol
PubMed ID:27857708
The marine ecosystem along the Western Antarctic Peninsula undergoes a dramatic seasonal transition every spring, from almost total darkness to almost continuous sunlight, resulting in a cascade of environmental changes, including phytoplankton blooms that support a highly productive food web. Despite having important implications for the movement of energy and ... More