Can the Qubit Flex Assay Tube Strips (Cat. No. Q33252) be cut to run 1 sample or does the full tube strip need to be placed in the Qubit Flex Fluorometer (Cat. No. Q33327)?
Qubit Flex Assay Tube Strips (Cat. No. Q33252) can be cut to run 1 sample with the Qubit Flex Fluorometer (Cat. No. Q33327).
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.