Qubit™ Flex System Verification Assay Kit - FAQs
Qubit™ Flex System Verification Assay Kit - FAQs
View additional product information for Qubit™ Flex System Verification Assay Kit - FAQs (Q33254)
1 product FAQs found
Does the Qubit Flex NGS Starter Kit (Cat. No. Q45893) include the Qubit Flex System Verification Assay Kit (Cat. No. Q33254)?
No. The Qubit Flex NGS Starter Kit (Cat. No. Q45893) does not include the Qubit Flex System Verification Assay Kit (Cat. No. Q33254). It must be purchased separately.
The description of the Qubit Flex Starter Packages (Cat. Nos. Q32894, Q45893, Q45894) can be found on the Qubit Flex Starter Packages product page.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.