Are Cat. Nos. Q33267, Q33265 and Q33266 all the same with the same buffer?
Quant-iT 1X dsDNA Assay Kits, high sensitivity (HS) and broad range (BR) is Cat. No. Q33267.
Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits is Cat. No. Q33265.
Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits is Cat. No. Q33266.
Cat. No. Q33267 is a Quant-iT kit, while the other two are exactly same version of Qubit, just two different sizes. The kits are very similar, but optimized for different applications, and they also have a different number of standards.
Quant-iT 1X dsDNA Assay Kit, broad range (BR) (Cat. No. Q33267)
- 1 x dsDNA, broad range 4 to 4000 ng
- 1000 (200 μL Assay Volume)
Qubit 1X dsDNA Broad Range (BR) Assay Kit (Cat. No. Q33265) - 1X dsDNA, broad range 4 to 4000 ng
- 100 Reactions
Qubit 1X dsDNA Broad Range (BR) Assay Kit (Cat. No. Q33266) - 1X dsDNA, broad range 4 to 4000 ng
- 500 Reactions
The Qubit 1X dsDNA BR Assay Kits are specifically designed to be used with Qubit fluorometers, particularly the Qubit 4 and Qubit Flex models.
The Quant-iT kits can be used with both Qubit fluorometers and microplate readers, making them more versatile in terms of equipment compatibility.
However QuantiT kit can be used on Qubit and vice versa. More information about kits can be found in the Application Note: Comparison of Quant-iT and Qubit DNA quantitation assays for accuracy and precision.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits is Cat. No. Q33265.
Qubit 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits is Cat. No. Q33266.
Cat. No. Q33267 is a Quant-iT kit, while the other two are exactly same version of Qubit, just two different sizes. The kits are very similar, but optimized for different applications, and they also have a different number of standards.
Quant-iT 1X dsDNA Assay Kit, broad range (BR) (Cat. No. Q33267)
- 1 x dsDNA, broad range 4 to 4000 ng
- 1000 (200 μL Assay Volume)
Qubit 1X dsDNA Broad Range (BR) Assay Kit (Cat. No. Q33265) - 1X dsDNA, broad range 4 to 4000 ng
- 100 Reactions
Qubit 1X dsDNA Broad Range (BR) Assay Kit (Cat. No. Q33266) - 1X dsDNA, broad range 4 to 4000 ng
- 500 Reactions
The Qubit 1X dsDNA BR Assay Kits are specifically designed to be used with Qubit fluorometers, particularly the Qubit 4 and Qubit Flex models.
The Quant-iT kits can be used with both Qubit fluorometers and microplate readers, making them more versatile in terms of equipment compatibility.
However QuantiT kit can be used on Qubit and vice versa. More information about kits can be found in the Application Note: Comparison of Quant-iT and Qubit DNA quantitation assays for accuracy and precision.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.