Search
View additional product information for imMedia™ Growth Medium, agar, kanamycin - FAQs (Q61120)
7 product FAQs found
The first is our formulation of LB agar (catalog number 22700-025), which has LB Media and agar mixed together. This comes in a 500g and 2.5 kg size. All you have to do is measure out, add water, and autoclave.
The second option is even easier. Our imMedia comes premeasured in pouches. We have three types: imMedia Amp Agar, Kan Agar and Zeo Agar for ampicillin, kanamycin and Zeocin-containing plates. All you have to do is empty the pouch, add water, heat and pour plates. No autoclaving is needed! Each pouch contains sufficient reagents for 8-10 plates.
imMedia Amp Agar, 20 pouches, Q601-20
imMedia Kan Agar, 20 pouches, Q611-20
imMedia Zeo Agar, 20 pouches, Q621-20
For added convenience, you should also check out our LB medium imMedia products: (Each pouch makes 200 mL of media)
imMedia Amp Liquid, 20 pouches, Q600-20
imMedia Kan Liquid, 20 pouches, Q610-20
imMedia Zeo Liquid, 20 pouches, Q620-20