QuantiGene™ Plex Plate Seals
Invitrogen™
QuantiGene™ Plex Plate Seals
QuantiGene Plex Plate Seals are clear microtiter plate seals for use with QuantiGene Plex magnetic separation plates (96- and 384-well).
Have Questions?
Catalog NumberQuantity
QG052445 seals
Catalog number QG0524
Price (USD)
-
Quantity:
45 seals
QuantiGene Plex Plate Seals are clear microtiter plate seals for use with QuantiGene Plex magnetic separation plates (96- and 384-well). These seals are intended to be used only during the branched DNA reagent hybridizations and SAPE incubation on the second day of the QuantiGene Plex Assay.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)QuantiGene Plex Magnetic Separation Plates
Product TypeAdhesive Seal
Quantity45 seals
Shipping ConditionRoom Temperature
ColorClear
Plate Compatibility96/384-well Magnetic Separation Plates
Product LineQuantiGene
Unit SizeEach
Contents & Storage
45 clear adhesive plate seals