QuantiGene™ Plex Human ADMETOX Panel, 80-plex
Invitrogen™

QuantiGene™ Plex Human ADMETOX Panel, 80-plex

The QuantiGene Plex Human ADMETOX Panel, 80-plex, enables the measurement of gene expression of all relevant ADME (absorption, distribution, metabolism,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
QGP-380-ADMETOX10 Plates
QGP-180-ADMETOX1 Plate
QGP-280-ADMETOX3 Plates
Catalog number QGP-380-ADMETOX
Price (USD)
86,700.00
Each
Quantity:
10 Plates
Price (USD)
86,700.00
Each

The QuantiGene Plex Human ADMETOX Panel, 80-plex, enables the measurement of gene expression of all relevant ADME (absorption, distribution, metabolism, and excretion) and toxicological biomarkers through the analysis of 80 RNA targets in a single well using Luminex xMAP technology in combination with branched DNA signal amplification. Analyze 80 genetic targets implicated in drug metabolism, including enzymes, transporters, and CYPs, as well as toxicology and apoptosis for efficient profiling of your drug discovery and basic research samples.

QuantiGene Plex panels are extensively tested for target combinability, interference, and cross-reactivity to provide the highest level of validation and precision. This kit is not compatible with the MAGPIX instrument.

This panel is used in combination with the QuantiGene Plex Gene Expression Assay Ordering.

Features of the QuantiGene Plex Human ADMETOX Panel, 80-plex, include:

  • True multiplexing—measure 80 genes of interest and housekeeping genes in the same well with no cross-reactivity
  • Accommodation of difficult sample types—works with degraded and cross-linked RNA in FFPE tissues and blood
  • Standardized platform—uses 96-well plate format compatible with Luminex 200, FLEXMAP 3D, and INTELLIFLEX systems (384-well format available on demand)
  • Simple workflow—employs an ELISA-like workflow with direct hybridization of transcripts to beads and transcript labeling (branched DNA signal amplification and PE readout)
  • Customize with validated targets—if alternative targets are needed in this panel, select from our large inventory of over 22,000 genes to QuantiGene Plex data analysis app.

    Applications

    QuantiGene Plex assay research applications include compound screening of drug candidates, biomarker verification, siRNA knockdown efficiency, prospective and/or retrospective analysis of clinical samples, microarray validation, predictive toxicology, and detection of translocations and fusion genes. In general, any multiplex gene expression analysis can be done with QuantiGene Plex assays, independent of species or sample type.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)QuantiGene Gene Expression Assay
For Use With (Equipment)Luminex Instrument
Product LineQuantiGene™ Plex
Sample TypePurified RNA, cell/blood/FFPE lysates, tissue homogenates
Shipping ConditionWet Ice
Detection MethodFluorescence
Format96-well
Product TypeMultiplex Panel
Quantity10 Plates
Research AreaToxicology
SpeciesHuman
Unit SizeEach
Contents & Storage

Target-specific probe set and magnetic capture beads (premixed and ready to use).

Store probe set and capture beads at 4°C.

Do not freeze the capture beads, as they can be damaged if frozen.