VetMAX™ Porcine PCV2 Quant Kit - FAQs
VetMAX™ Porcine PCV2 Quant Kit - FAQs
View additional product information for VetMAX™ Porcine PCV2 Quant Kit - FAQs (QPCV)
2 product FAQs found
What are the probes included in the VetMAX Porcine PCV2 Quant Kit?
The TaqMan assay for QPCV2 is labeled with FAM and quenched by NFQ (non-fluorescent quencher). The TaqMan assay for IPC is labeled with VIC and quenched with TAMRA.
Is the qPCR master mix included with the VetMAX Porcine PCV2 Quant Kit?
Yes, the qPCR master mix is supplied in the Green tube (3-Mix QPCV2).