Invitrogen™

QuantiGene™ Singleplex Assay Kit

Catalog number:  QS0011

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

QuantiGene™ Singleplex Assays are hybridization-based assays for the direct quantitation of RNA, DNA, or microRNA transcripts directly from cell lysates or tissue homogenates. They utilize branched DNA technology for signal amplification and signal is detected using a standard luminometer. The QuantiGene Singleplex Assay Kit contains all buffers, plates, and detection reagents necessary to perform the assay in 96-well plates.

All capture plates in this assay kit are packaged together in a single container, and are NOT packaged in individual pouches.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Size: 50 x 96-well plates

Contents & storage

Blocking Reagent (10 mL, store at -20°C)
Pre-Amplifier (550 µL, store at -20°C)
Amplifier (550 µL, store at -20°C)
Capture Plate (50 ea, store at 2-8°C)
Label Probe (550 µL, store at 2-8°C)
Substrate (530 mL, store at 2-8°C)
Amplifier/Label Probe Diluent (1800 mL, store at 15-30°C)
Lysis Mixture (410 mL, store at 15-30°C)
Wash Buffer Component 1 (75 mL, store at 15-30°C)
Wash Buffer Component 2 (125 mL, store at 15-30°C)
Plate Seals (260 ea, store at 15-30°C)

Documents

Manuals & protocols