Search
| 製品番号(カタログ番号) | 数量 |
|---|---|
| QS0506 | 12 Seals |
| QS0507 | 260 Seals |
QuantiGene Singleplex Foil Plate Seals are intended for use with all hybridization steps of the QuantiGene Singleplex Assay. The foil plate seals are compatible with 96- and 384-well plates.