QuantiGene™ Singleplex Foil Plate Seals
Invitrogen™
QuantiGene™ Singleplex Foil Plate Seals
QuantiGene Singleplex Foil Plate Seals are intended for use with all hybridization steps of the QuantiGene Singleplex Assay.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
QS0507260 sellos
QS050612 sellos
Número de catálogo QS0507
Precio (MXN)
-
Cantidad:
260 sellos
QuantiGene Singleplex Foil Plate Seals are intended for use with all hybridization steps of the QuantiGene Singleplex Assay. The foil plate seals are compatible with 96- and 384-well plates.
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Tipo de productoPelícula, cierre o aplicador
Cantidad260 sellos
Condiciones de envíoTemperatura ambiente
N.° de pocillos96
Plate CompatibilityMicroplaca
Línea de productosQuantiGene
Unit SizeEach
Contenido y almacenamiento
260 sellos de placa adhesiva con dorso de papel de aluminio