ViewRNA™ Chromogenic Signal Amplification Kit (1-plex) - FAQs
ViewRNA™ Chromogenic Signal Amplification Kit (1-plex) - FAQs
View additional product information for ViewRNA™ Chromogenic Signal Amplification Kit (1-plex) - FAQs (QVT0200)
1 product FAQs found
Do you have an alternative for the ViewRNA ISH Tissue Assay Kit (2-plex) and ViewRNA Chromogenic Signal Amplification (1-plex) Kits (Cat. Nos. QVT0012, QT0200)?
Unfortunately, there is no exact comparable kit to the discontinued kit ViewRNA Chromogenic Signal Amplification Kit (1-plex) (Cat. No. QVT0200) as the use of the Naphthol Buffer is no longer required for signal amplification.
The closest replacement we currently offer is the ViewRNA Colorimetric Tissue Core Kit and Individual Modules:
ViewRNA Colorimetric Tissue Core Kit and Individual Modules (Cat. No. QVT0400)