ViewRNA™ Fast Red Module, 24 Assays - FAQs
ViewRNA™ Fast Red Module, 24 Assays - FAQs
View additional product information for ViewRNA™ Colorimetric Tissue Core Kit and Individual Modules - FAQs (QVT0400A, QVT0400B, QVT0400, QVT0410, QVT0420, QVT0430)
2 product FAQs found
Do you offer an alternative for the discontinued ViewRNA ISH Tissue Assay kits (2-plex) (Cat. No. QVT0012 and QVT0013)?
Currently, we offer the ViewRNA Colorimetric Tissue Assays in three formats:
- ViewRNA Colorimetric Tissue Core Kit (Cat. No. QVT0400) and  Individual modules for Fast Red (Cat. No. QVT0410), Fast Blue (Cat. No. QVT0420) and DAB (Cat. No. QVT0430) signal development
- ViewRNA Colorimetric Tissue 1-Plex Kits (Cat. No. QVT0420C, QVT4420C, QVT0410C, QVT4410C, QVT0430C, QVT4430C)
- ViewRNA Colorimetric Tissue 2-Plex Kits (Cat. No. QVT0433C, QVT4433C, QVT0412C, QVT4412C, QVT0422C, QVT4422C)
Do you have an alternative for the ViewRNA ISH Tissue Assay Kit (2-plex) and ViewRNA Chromogenic Signal Amplification (1-plex) Kits (Cat. Nos. QVT0012, QT0200)?
Unfortunately, there is no exact comparable kit to the discontinued kit ViewRNA Chromogenic Signal Amplification Kit (1-plex) (Cat. No. QVT0200) as the use of the Naphthol Buffer is no longer required for signal amplification.
The closest replacement we currently offer is the ViewRNA Colorimetric Tissue Core Kit and Individual Modules:
ViewRNA Colorimetric Tissue Core Kit and Individual Modules (Cat. No. QVT0400)