ViewRNA™ Blue Brown 2-Plex Kit, 24 Assays - FAQs
ViewRNA™ Blue Brown 2-Plex Kit, 24 Assays - FAQs
View additional product information for ViewRNA™ Colorimetric Tissue 2-Plex Kits - FAQs (QVT4412C, QVT0412C, QVT4433C, QVT4422C, QVT0433C, QVT0422C)
1 product FAQs found
Do you offer an alternative for the discontinued ViewRNA ISH Tissue Assay kits (2-plex) (Cat. No. QVT0012 and QVT0013)?
Currently, we offer the ViewRNA Colorimetric Tissue Assays in three formats:
- ViewRNA Colorimetric Tissue Core Kit (Cat. No. QVT0400) and  Individual modules for Fast Red (Cat. No. QVT0410), Fast Blue (Cat. No. QVT0420) and DAB (Cat. No. QVT0430) signal development
- ViewRNA Colorimetric Tissue 1-Plex Kits (Cat. No. QVT0420C, QVT4420C, QVT0410C, QVT4410C, QVT0430C, QVT4430C)
- ViewRNA Colorimetric Tissue 2-Plex Kits (Cat. No. QVT0433C, QVT4433C, QVT0412C, QVT4412C, QVT0422C, QVT4422C)