ViewRNA™ Tissue Positive Control Kit (1-plex) (4 rat kidney slides + probe) - FAQs
ViewRNA™ Tissue Positive Control Kit (1-plex) (4 rat kidney slides + probe) - FAQs
View additional product information for ViewRNA™ Tissue Positive Control Kit (1-plex) (4 rat kidney slides + probe) - FAQs (QVT0519)
1 product FAQs found
Do you offer any positive controls for the ViewRNA tissue assay kits?
Yes, you can use the ViewRNA Tissue Positive Control Kit (1-plex) (Cat. No. QVT0519) or the ViewRNA Tissue Positive Control Kit (2-plex) (4 rat kidney slides + probes) (Cat. No. QVT0516). The positive control includes four each of 5 µm thick rat kidney FFPE sections attached to positively charged slides as well as one (1-plex) or two (2-plex) probe sets for housekeeping genes.