Fluorescein-12-dUTP Solution (1 mM)
Thermo Scientific™

Fluorescein-12-dUTP Solution (1 mM)

Thermo Scientific Fluorescein-12-dUTP (fluorescein-5(6)-carboxaminocaproyl-[5-{3-aminoallyl}-2'-deoxyuridine-5-triphosphate]) is supplied as 1 mM aqueous solution.Fluorescein-12-dUTP can be enzymatically incorporated into DNA with:• Reverse transcriptases•Read more
Have Questions?
Catalog NumberQuantity
R010125 μL
Catalog number R0101
Price (USD)
223.65
Online exclusive
269.00
Save 45.35
Each
Quantity:
25 μL
Request bulk or custom format
Price (USD)
223.65
Online exclusive
269.00
Save 45.35
Each

Thermo Scientific Fluorescein-12-dUTP (fluorescein-5(6)-carboxaminocaproyl-[5-{3-aminoallyl}-2'-deoxyuridine-5-triphosphate]) is supplied as 1 mM aqueous solution.

Fluorescein-12-dUTP can be enzymatically incorporated into DNA with:
• Reverse transcriptases
• Taq DNA polymerase
• phi29 DNA Polymerase
• Klenow Fragment, exo-
• Klenow Fragment
• DNA Polymerase I

Applications
• Enzymatic non-radioactive labeling of DNA during cDNA synthesis
• PCR
• Nick-translation
• Random primed labeling
• Primer extension

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Labeling MethodDirect Labeling
PolymeraseDNA Polymerase
Quantity25 μL
Shipping ConditionDry Ice
Concentration1 mM
For Use With (Application)cDNA Synthesis, Non-Radioactive Labeling
FormLiquid
Label or DyeFluorescein (FITC)
Unit SizeEach
Contents & Storage
Fluorescein-12-dUTP Solution, 1 mM (25 μL)

Store at –20°C.