Product Image
Thermo Scientific™

Anaerobic Reducible LKV Blood Agar

Cultivate and isolate fastidious anaerobic bacteria with this selective, ready-to-use solid medium.
Have Questions?
Catalog NumberFormat
R0106812mm x 85mm Monoplate
Catalog number R01068
Price (USD)
-
Format:
12mm x 85mm Monoplate

Cultivate and isolate fastidious anaerobic bacteria with Thermo Scientific™ Remel™ Anaerobic Reducible LKV Blood Agar. Adding reducing agents lowers the oxidation-reduction potential of the medium and improves the recovery of anaerobic organisms1. Kanamycin and vancomycin are added as selective agents for recovery of gram-negative anaerobes, especially Bacteriodes spp2.

Anaerobic Reducible Blood Agar is a solid media recommended for use in qualitative procedures for the primary isolation and cultivation of fastidious anaerobic bacteria.

  • Selective: Kanamycin and vancomycin antibiotics inhibit gram-positive organisms and facultative anaerobic bacteria.
  • Easy-to-detect: Laked blood is added to enhance pigment formation by anaerobic bacteria.
  • Ready-to-use: Convenience of prepared media.

Anaerobic Reducible Blood Agar is a nutritious medium containing peptone, hemin, vitamin K and sheep blood, which supply anaerobic organisms with required growth factors. Colistin and nalidixic inhibit the growth of most gram-negative bacilli. Kanamycin and vancomycin inhibit gram-positive organisms and facultative anaerobic bacteria. Laked blood is added to enhance pigment formation by anaerobic bacteria.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

General References:

  1. Ellner, P.D., Granato, P.A. and May, C.B. 1973. Appl. Microbiol. 26: 904–913.
  2. Finegold, S.M. 1965. International Mikrobiologisches Symposium Potsdam, Germany, Ehrnahrungsforchung. 10:517–528
Specifications
DescriptionAnaerobic Reducible LKV Blood Agar
Format12mm x 85mm Monoplate
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach