Remel™ BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide
Thermo Scientific™
Remel™ BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide
Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide is used for selective isolation of fastidious fungi and yeast.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatPackaging Type
R0114810/Pk.Monoplate Deep Pour10/Pk.
R08078100/Pk.F - Slants100/Pk.
Catalog number R01148
Price (USD)
-
Quantity:
10/Pk.
Format:
Monoplate Deep Pour
Packaging Type:
10/Pk.
Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide is used for selective isolation of fastidious fungi and yeast.
  • Chloramphenicol inhibits some gram-positive and gram-negative microorganisms
  • Gentamicin inhibits some gram-negative bacteria
  • Cycloheximide inhibits rapid growing saprophytic fungi
Specifications
DescriptionBHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide
FormatMonoplate Deep Pour
Packaging Type10/Pk.
Quantity10/Pk.
Product TypeAgar
Unit SizeEach