Product Image
Thermo Scientific™
Remel™ BHI Agar w/Sheep Blood, Penicillin, Streptomycin, Yeast Extract
Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Penicillin, Streptomycin, Yeast Extract is for selective isolation of fastidious fungi and yeast.
Have Questions?
Catalog NumberdimFormat
R01160
Catalog number R01160
Price (USD)
-
Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Penicillin, Streptomycin, Yeast Extract is for selective isolation of fastidious fungi and yeast.
  • Yeast extract acts as a growth stimulant
  • Penicillin inhibits gram-positive bacteria
  • Streptomycin inhibits some gram-positive and gram-negative bacteria
Specifications
DescriptionBHI Agar w/Sheep Blood, Penicillin, Streptomycin, Yeast Extract
FormatMonoplate Deep Pour
Quantity10/Pk.
Product TypeAgar
Unit SizeEach