Remel™ BiGGY Agar
Thermo Scientific™
Remel™ BiGGY Agar
Use for isolation and presumptive identification of Candida spp.
Have Questions?
Catalog NumberFormat
R0118012mm x 85mm Monoplate
Catalog number R01180
Price (USD)
-
Format:
12mm x 85mm Monoplate
Thermo Scientific™ Remel BiGGY Agar (Bismuth Sulfite Glucose Glycine Yeast) (Nickerson Agar) is for isolation and presumptive identification of Candida spp.
See also CandidaTube™
Specifications
DescriptionBiGGY Agar (Bismuth Sulfite Glucose Glycine Yeast) (Nickerson Agar)
Format12mm x 85mm Monoplate
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach