Bile Esculin Agar
Thermo Scientific™
Bile Esculin Agar
Isolate and differentiate enterococci / Group D streptococci and non-Group D streptococci using this selective microbiological medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R0119010/Pk.
R06029420/Pk.
Catalog number R01190
Price (USD)
-
Quantity:
10/Pk.
Isolate and differentiate enterococci/Group D streptococci and non Group D streptococci using Thermo Scientific™ Bile Esculin Agar, a selective microbiological medium.
Enterococci / Group D streptococci hydrolyse aesculin to form aesculetin and dextrose. Aesculetin combines with ferric citrate in the medium to form a dark brown or black complex which is indicative of a positive result. Bile salts will inhibit Gram-positive bacteria other than enterococci / Group D streptococci.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Specifications
FormatMonoplate
Media TypeBile Esculin Agar
Quantity10/Pk.
Manufacturing OriginUnited States
Product TypeAgar Plate
Unit SizeEach