Bile Esculin Azide Agar
Thermo Scientific™

Bile Esculin Azide Agar

Isolate, enumerate and presumptively identify group D streptococci and enterococci with a medium that utilizes bile salts to improve selectivity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatMedia Type
R0119410/Pk.12mm x 85mm MonoplateBile Esculin Azide Agar
R01189100/Pk.12mm x 85mm MonoplateBile Esculin Azide Agar
Catalog number R01194
Price (USD)
-
Quantity:
10/Pk.
Format:
12mm x 85mm Monoplate
Media Type:
Bile Esculin Azide Agar
Isolate, enumerate and presumptively identify group D streptococci and enterococci using Thermo Scientific™ Bile Esculin Azide Agar, a microbiological media that includes a high portion of Ox bile.
Ox bile present in the media inhibits the growth of Gram-positive bacteria except Enterococci, while Sodium azide suppresses the Gram-negative bacteria. Group D streptococci and enterococci are able to hydrolyse aesculin into aesculetin and glucose, and the bondage of aesculetin and ferric ammonium citrate causes the brown-black to black halo around the colonies.

Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery we recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Certifications/ComplianceIndustrial Reference: BAM, SMWW.
Format12mm x 85mm Monoplate
Media TypeBile Esculin Azide Agar
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach