Columbia Agar with Sheep Blood PLUS
Columbia Agar with Sheep Blood PLUS
Columbia Agar with Sheep Blood PLUS
Thermo Scientific™

Columbia Agar with Sheep Blood PLUS

Isolate and cultivate fastidious microorganisms with clearly visible haemolytic reactions (staphylococci and streptococci) with a medium containing sheep blood.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatMedia TypeManufacturing Origin
R01218150/Pk.10mm x 100mm Spacesaver PlateBlood Agar with 5% Sheep BloodUnited States
R0121510/Pk.85mm MonoplateBlood Agar with 5% Sheep BloodUnited States
R01217100/Pk.85mm MonoplateBlood Agar with 5% Sheep BloodUnited States
Catalog number R01218
Price (USD)
-
Quantity:
150/Pk.
Format:
10mm x 100mm Spacesaver Plate
Media Type:
Blood Agar with 5% Sheep Blood
Manufacturing Origin:
United States
Isolate and cultivate fastidious microorganisms with clearly visible haemolytic reactions (Staphylococci and Streptococci) with a media formulation that contains sheep blood. Thermo Scientific™ Columbia Agar with Sheep Blood PLUS promotes haemolysis typical of Staphylococcus aureus, and it gives typical growth for Streptococcus pneumonia (dent morphology).
Grow fastidious organisms with rapid production of large colonies, clearly defined zones of haemolysis and good colonial differentiation for all-around performance. Traditionally, blood agar has been either casein hydrolysate or meat infusion media. The advantage of the first lies in the rapid production of large colonies, and the second in clearly defined zones of haemolysis and good colonial differentiation. This formulation combines the virtues of both for optimal performance.

Staphylococcus aureus
is a coagulase-positive, catalase positive, gram-positive cocci that occurs singly or in pairs, tetrads, short chains, and grape-like clusters. It is commonly carried on the skin and in the nose of healthy individuals but is also a major cause of infection in both healthcare and community settings worldwide. S. aureus can produce a variety of syndromes with clinical manifestations including skin and soft tissue infections, empyema, bloodstream infections, pneumonia, osteomyelitis, septic arthritis, endocarditis, sepsis, and meningitis.

Streptococci are facultatively anaerobic, catalase-negative, gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fascitis. One of the most useful phenotypic characteristics of streptococci is the hemolytic reaction, generally classified as alpha, beta or gamma according to the appearance of zones around colonies growing on blood agar. β-hemolytic streptococci are further characterized by Lancefield groups based on the antigenic differences in group-specific polysaccharides located in the bacterial cell wall.

Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery we recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceIndustrial Reference: IDF.
Format10mm x 100mm Spacesaver Plate
Media TypeBlood Agar with 5% Sheep Blood
Quantity150/Pk.
Manufacturing OriginUnited States
Product TypeAgar Plate
Unit SizeEach