Product Image
Thermo Scientific™
Remel™ Blood Agar w/Neomycin 30μg
Use for selective isolation of Streptococcus pyogenes and Streptococcus pneumoniae.
Have Questions?
Catalog NumberQuantity
R0122830 μg
Catalog number R01228
Price (USD)
-
Quantity:
30 μg
The Thermo Scientific™ Remel Blood Agar w/Neomycin 30μg (TSA w/5% Sheep Blood) is for selective isolation of Streptococcus pyogenes and Streptococcus pneumoniae.
  • Neomycin inhibits Enterobacteriaceae
Specifications
DescriptionBlood Agar w/Neomycin (TSA w/5% Sheep Blood)
Format12mm x 85mm Monoplate
Quantity30 μg
Product TypeAgar Plate
Unit SizeEach