Brilliant  Green Agar
Thermo Scientific™

Brilliant Green Agar

Selectively isolate and differentiate Salmonella species other than Salmonella typhi from food and other materials with this selective medium.

Have Questions?
Catalog NumberFormat
R0124612mm x 85mm Monoplate
Catalog number R01246
Price (USD)
-
Format:
12mm x 85mm Monoplate

Selectively isolate and differentiate Salmonella species other than Salmonella typhi from food and other materials with Thermo Scientific™ Remel™ Brilliant Green Agar. The medium is recommended by the American Public Health (APHA) for the recovery of Salmonella from food products1.

Salmonella infection (salmonellosis) is a common bacterial disease that affects the intestinal tract. Salmonella bacteria typically live in animal and human intestines and are shed through feces. Humans become infected most frequently through contaminated water or food.

Use Brilliant Green Agar for isolation of salmonellae other than Salmonella typhi.

  • Selective: brilliant green in the medium inhibits the growth of non-target organisms.
  • Easy-to-read: Salmonella colonies are easy to read as they range from reddish to pink to nearly white in color with red zone.
  • Corresponds to the medium recommended by the APHA1

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

Not intended for IVD use.
1. Downes, F.p. and K.Ito.2001. Compendium of Methods for the Microbiological Examination of foods 4th Ed. APHA, Washington, D.C.
Specifications
Certifications/ComplianceIndustrial Reference: SMWW, USDA, USP.
DescriptionBrilliant Green Agar
Format12mm x 85mm Monoplate
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach