Remel™ CCFA Modified Agar w/Horse Blood (with cefoxitin, cycloserine, fructose)
Thermo Scientific™
Remel™ CCFA Modified Agar w/Horse Blood (with cefoxitin, cycloserine, fructose)
Use for selective isolation and differentiation of Clostridium difficile.
Have Questions?
Catalog NumberFormat
R0126612mm x 85mm Monoplate
Catalog number R01266
Price (USD)
-
Format:
12mm x 85mm Monoplate
Thermo Scientific™ Remel CCFA Modified Agar w/Horse Blood (with cefoxitin, cycloserine, fructose) is used for selective isolation and differentiation of Clostridium difficile.
Specifications
DescriptionCCFA Modified Agar w/Horse Blood w/cefoxitin, cycloserine, fructose
Format12mm x 85mm Monoplate
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach