Candida BCG Agar
Thermo Scientific™
Candida BCG Agar
Selectively isolate and presumptively identify Candida species with this selectve ready prepared medium.
Have Questions?
Catalog NumberdimFormat
R01282
Catalog number R01282
Price (USD)
-
Selectively isolate and presumptively identify Candida species with Thermo Scientific™ Remel™ Candida BCG Agar. Candida species ferment the dextrose in the medium causing a reduction in pH in the agar. Bromocresol green indicator changes from green to yellow when dextrose in fermented causing Candida species to form yellow to yellow-green colonies with a yellow zone.
The Candida BCG Agar is recommended in the Laboratory Methods in Medical Mycology for the isolation and presumptive identification of Candida spp on the basis of colony morphology1.
Serious infections due to Candida species are becoming increasingly prevalent. This poses particular problems because of the increasing incidence of non-albicans spp. and the emergence of non-albicans isolates resistant to both amphotericin B and the newer azoles.2
Use Candida BCG Agar for selective isolation and presumptive identification of Candida species.
  • Selective: selective agent neomycin inhibits aerobic and facultative anaerobic Gram-negative bacteria and some Gram-positive species making it selective for Candida
  • Easy-to-read: Candida species are easy to read as they form yellow to yellow-green colonies with a yellow zone.
  • Ready-to-use: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
General References:
  1. Haley, L.D. and C.S. Callaway. 1978. Laboratory Mathods in Medical Mycology. 4th ed. U.S. Dept of H.H.S.CDC, Atlanta, GA.
  2. Sheehan, D. J. et al. (1999) Current and Emerging Azole Antifungal Agents. Clinical Microbiology Reviews, 12(1): 40-79.
Specifications
DescriptionCandida BCG Agar
FormatMonoplate Deep Pour
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach