Provides selective and differential isolation of Cryptococcus gattii (i.e. serotypes B and C) from other Cryptococcus spp.
Have Questions?
Catalog Number
Format
R01299
12mm x 85mm Monoplate
Catalog number R01299
Price (USD)
-
Format:
12mm x 85mm Monoplate
Thermo Scientific™ Remel CGB Agar provides selective and differential isolation of Cryptococcus gattii (i.e. serotypes B and C) from other Cryptococcus spp.