Haemophilus Isolation Agar w/bacitracin and horse blood
Thermo Scientific™
Haemophilus Isolation Agar w/bacitracin and horse blood
Cultivate and isolate fastidious anaerobic bacteria with this selective, ready-to-use solid medium.
Have Questions?
Catalog NumberQuantity
R0147010/Pk.
Catalog number R01470
Price (USD)
-
Quantity:
10/Pk.
Selectively isolate Haemophilus species from respiratory specimens with Thermo Scientific™ Remel™ Haemophilus Isolation Agar w/bacitracin and horse blood. Haemophilus influenzae, a fastidious gram-negative bacillus, is a common cause of serious diseases of the upper and lower respiratory tract1.
Haemophilus Isolation Agar w/bacitracin and horse blood is a solid media recommended for use in qualitative procedures for selective isolation of Haemophilus spp. from respiratory specimens.
  • Selective: Kanamycin and vancomycin antibiotics inhibit gram-positive organisms and facultative anaerobic bacteria. Neomycin stimulates the growth of gram-negative anaerobes, especially Bacteriodes spp. Colistin and nalidixic inhibit the growth of most gram-negative bacilli.
  • Enriched medium: The presence of intact erythrocytes in horse blood enables differentiation of hemolytic and nonhemolytic Haemophilus spp. Horse blood, unlike sheep blood, does not contain V factor-inactivating enzymes.
  • Ready-to-use: Convenience of prepared media.
Beef heart Infusion and casein peptone supply the nutritional requirements for all but the fastidious bacteria. Horse sheep supplies both the X and V factors, which are growth requirements for certain organisms, such as Haemophilus influenzae. Horse blood also serves to distinguish the hemolytic species of Haemophilus from non-hemolytic Haemophilus species. Bacitracin inhibits normal flora found in respiratory cultures, particularly Neisseria spp.4.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
General References:
  1. Chapin, K.C and Doern, G.V. 1983. J. Clin. Microbiol. 17:1163–1165.
  2. Klein, M. and Blazevic, D.J. 1970. Am. J. Med. Tech. 36:97–106.
  3. Koneman, E.W., Allen, S.D., Janda, W.M., Schreckenberger, P.C. and Winn, W.C, Jr. 1997. Color Atlas and Textbook of Diagnostic Microbiology. 5th ed. Lippincott Williams and Wilkins, Philadelphia, PA.
  4. Hovig, B and Aandahl, E.H. 1969. Acta. Pathol. Microbiol. Scand. 77:676–684.
Specifications
DescriptionHaemophilus Isolation Agar w/bacitracin, horse blood
Format12mm x 85mm Monoplate
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach