HBT Bilayer Medium
Thermo Scientific™
HBT Bilayer Medium
Selectively isolate and presumptively identify Gardnerella vaginalis using HBT Bilayer Medium (Human Blood-Polysorbate 80).
Have Questions?
Catalog NumberFormat
R0147812mm x 85mm Monoplate
Catalog number R01478
Price (USD)
-
Format:
12mm x 85mm Monoplate
Selectively isolate and presumptively identify Gardnerella vaginalis using Thermo Scientific™ Remel™ HBT Bilayer Medium (Human Blood-Polysorbate 80). Media used by earlier investigators to recover G. vaginalis from clinical specimens include chocolate agar, peptone starch dextrose (PSD) agar, and Columbia colistin nalidixic acid agar1-3. Greenwood and Pickett developed V agar and demonstrated that G. vaginalis is a beta-hemolytic on agar containing human blood but not on sheep blood agar4,5.
Gardnerella vaginalis is a gram-variable rod or coccobacillus. It is a venereally transmitted bacterium that causes bacterial vaginosis6. This is a condition characterized by an increase in the numbers of G. vaginalis and various obligate anaerobes, and a concomitant decrease in lactobacilli observed on gram-stained smears. HBT Bilayer Medium, formulated by Totten et al., consists of a bottom layer of Columbia colistin nalidixic acid agar supplemented with peptone, amphotericin B, and polysorbate 80 together with a top layer of the same media with 5% human blood added7. It shows significantly higher isolation rates of G. vaginalis than those obtained with other media.
Use HBT BiLayer Medium for selective and presumptive identification of Gardnerella vaginalis.
  • Ready to use – Convenience of prepared media
  • Selective medium - Colistin, nalidixic acid, and amphotericin B are selective agents that inhibit most gram-negative rods, yeast, and diphtheroids
  • Easy to interpret - G. vaginalis produces small, opaque colonies surrounded by a diffuse zone of beta-hemolysis
The casein and meat peptones supply nutrients. Phosphate buffer maintains pH and cornstarch neutralizes toxic fatty acids. Polysorbate 80 enhances the diffusion of beta-hemolysis. The top layer contains the same medium with addition of 5% human blood. Since the zones of beta-hemolysis are viewed through a shallow layer of blood-containing medium, hemolysis is more distinct and easier to detect than with a single layer medium.
General References:
  1. Dunkelberg, W.E., Jr., and R.I. Bosman. 1961. Mil.Med. 126:920-922.
  2. Dunkelberg, W.E., Jr., R. Skaggs, and D.S. Kellogg, Jr. 1970. Appl. Microbiol. 19:47-52.
  3. Golberg, R.L. and J.A. Washington. 1976. J. Clin. Microbiol. 4:245-247.
  4. Greenwood, J.R. and M.J. Pickett. 1979. J. Clin. Microbiol. 9:200-204.
  5. Piot, P., E. Van Dyck, P.A. Totten and K.K. Holmes. 1982. J. Clin. Microbiol. 15:19-24.
  6. Winn, W.C., Jr., S.D. Allen, W.M. Janda, E.W. Koneman, G.W. Procop, P.C. Schreckenberger, and G.L. Woods. 2006. Koneman's Color Atlas and Textbook of Diagnostic Microbiology. 6th ed. Lippincott Williams & Wilkins, Philadelphia, PA.
  7. Totten, P.A., R. A. Amsell, J.Hale, P.Piot, and K.K. Holmes. 1982. J. Clin. Microbiol. 15:141-147.
Specifications
DescriptionHBT Bilayer Medium
Format12mm x 85mm Monoplate
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach