Product Image
Thermo Scientific™
Inhibitory Mold Agar with Gentamicin and Chloramphenicol
Isolate and cultivate Histoplasma capsulatum, dermatophytes, and other pathogenic fungi with this selective, ready-to-use medium.
Have Questions?
Catalog NumberdimFormat
R01506
Catalog number R01506
Price (USD)
-
Isolate and cultivate Histoplasma capsulatum, dermatophytes, and other pathogenic fungi with Thermo Scientific™ Remel™ Inhibitory Mold Agar with Gentamicin and Chloramphenicol. Inhibitory Mold Agar was formulated by Ulrich as a general purpose medium for cultivation of fungi1. It is also recommended for selective isolation of pathogenic yeasts and molds (e.g. Cryptococcus neoformans, Histoplasma capulatum, and Trichophyton spp) from clinical specimens contaminated with commensal microbial flora2, 3.
Use Inhibitory Mold Agar with Gentamicin and Chloramphenicol for isolation and cultivation of Histoplasma capsulatum, dermatophytes, and other pathogenic fungi.
  • Selective: gentamicin and chloramphenicol act as selective agents. Chloramphenicol is a broad spectrum antibiotic which inhibits many Gram-positive and Gram-negative bacteria while gentamicin inhibits commensal microbial flora.
  • Ready-to-use: convenience of prepared media.
Casein and meat peptones supply nitrogenous compounds, amino acids, and peptides necessary for the growth of fungi. Dextrose, starch, and dextrin are ready sources of energy. Sodium chloride and metallic salts are essential electrolytes and help to maintain osmotic equilibrium. Yeast extract is a source of vitamins.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
General References:
  1. Ulrich, J.A. 1956. Abstract M75. Proceed. 56th Gen.Meet, Soc, Am. Bacteriol. Pages 87
  2. Haley, L.D. and C.S. Callaway. 1987. Laboratory Methods in Medical Mycology, U.S. Dept. of H.H.S. and CDC, Atlanta, GA
  3. Larone, D.H. 2002. Medically important fungi, A guide to Identification. 4th ed. ASM Press, Washington D.C.
Specifications
DescriptionInhibitory Mold Agar w/Gentamicin
FormatMonoplate Deep Pour
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach