Product Image
Thermo Scientific™
MRS Agar
Isolate and enumerate Lactobacillus species and gram-positive cocci using MRS Agar.
Have Questions?
Catalog NumberQuantity
R0158510/Pk.
Catalog number R01585
Price (USD)
-
Quantity:
10/Pk.
Isolate and enumerate Lactobacillus species and gram-positive cocci from clinical and non-clinical samples using Thermo Scientific™ Remel™ MRS Agar. In 1960, De Man, Rogosa and Sharpe modified a medium earlier formulated by Rogosa et al1 by eliminating tomato juice and created a medium which supported the growth of slower growing lactobacilli2. Recently MRS Agar has been used in the clinical laboratory to differentiate certain strains of gram-positive cocci and lactobacilli from clinical samples3-6.
‘Lactic acid bacteria’ are Gram-positive, catalase and oxidase negative and fastidious in their nutritional requirements. Growth is enhanced considerably by microaerobic conditions. Generally, the ‘lactic acid bacteria’ show delayed growth and smaller colony size than other microorganisms. They may be overgrown in non-selective media, especially if incubation is required for 2-4 days.
  • Can be used to differentiate certain strains of gram-positive cocci and lactobacilli.
  • Supports the growth of slower growing lactobacilli.
  • Selective: Ammonium citrate and sodium acetate inhibit the growth of commensal microbial flora, gram-negative bacilli and fungi, and improve the growth of lactobacilli.
  • Ready-to-use: Convenience of prepared media.
This medium contains gelatin peptone and beef extract that supply essential nutrients and amino acids necessary for bacterial growth. Yeast extract provides vitamin B complex and dextrose is the fermentable carbohydrate and energy source. Polysorbate 80 supplies fatty acids required for the metabolism of lactobacilli. Magnesium sulphate and manganese sulphate provide essential ions for multiplication of lactobacilli. Phosphates provide good buffering action in the media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
  1. Rogosa, M., R.F. Wiseman, J.A. Mitchell, M.N. Disraely, and A.J. Beaman. 1953. J. Bacteriol. 65:681-699.
  2. DeMan J.C., M. Rogosa, and M.E. Sharpe. 1960, J. Appl. Bacteriol., 23:130-135.
  3. Facklam, R., D. Hollis, and M.D. Collins. 1989. J. Clin. Microbiol. 27:724-730.
  4. Spitzer, E.D. 1990. Lab. Med. 21:411-413.
  5. Golledge, C.L. 1989. Clin. Microbiol. News. 11:29-30.
  6. Adams, S. 1991. Clin. Lab. Science. 4:93-94.
Specifications
Certifications/ComplianceIndustrial Reference: COMPF, IDF.
DescriptionMRS Agar (Man, Rogosa, Sharpe Agar)
FormatMonoplate
Quantity10/Pk.
Unit SizeEach