Product Image
Thermo Scientific™
Remel™ McCarthy Agar
Thermo Scientific™ Remel McCarthy Agar is for the isolation and differentiation of Gardnerella vaginalis.
Have Questions?
Catalog NumberFormat
R0159012mm x 85mm Monoplate
Catalog number R01590
Price (USD)
-
Format:
12mm x 85mm Monoplate
Thermo Scientific™ Remel McCarthy Agar is for the isolation and differentiation of Gardnerella vaginalis.
Specifications
DescriptionMcCarthy Agar
Format12mm x 85mm Monoplate
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach