Search
Isolate Burkholderia cepacia from respiratory secretions of patients with cystic fibrosis with these prepared plates.
| Catalog Number | dimFormat |
|---|---|
| R01653 |
Isolate Burkholderia cepacia from respiratory secretions of patients with cystic fibrosis with Thermo Scientific™ Remel™ OFPBL Medium.
Burkholderia cepacia grows slowly and exhibit variable colony morphology on commonly used media for the culture of respiratory specimens. As a result, it is easily obscured by overgrowth with Pseudomonas aeruginosa, other respiratory pathogens, and /or commensal microbial flora. OFPBL (oxidative-fermentative polymyxin B-bacitracin-lactose) medium is a selective and differential medium for the isolation of B. cepacia complex from respiratory specimens of patients with CF.
Burkholderia cepacia is a motile aerobic oxidase positive Gram-negative bacillus commonly found in liquid reservoirs and moist environments. It is an important opportunistic pathogen and causes pulmonary infection among individuals with cystic fibrosis (CF).
OFPBL medium is a selective and differential medium for isolation of B. cepacia complex from respiratory specimens of patients with CF.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.