Spectra™ MRSA Medium
Spectra™ MRSA Medium
Thermo Scientific™
Spectra™ MRSA Medium
Obtain fast and accurate results with a selective chromogenic medium for screening clinical samples for the presence of Methicillin Resistant Staphylococcus aureus.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatMedia TypeManufacturing Origin
R01822100/Pk.85mm MonoplateSpectra MRSA MediumUnited States
R0182110/Pk.85mm MonoplateSpectra MRSA MediumUnited States
Catalog number R01822
Price (USD)
-
Request A Quote
Quantity:
100/Pk.
Format:
85mm Monoplate
Media Type:
Spectra MRSA Medium
Manufacturing Origin:
United States
Obtain faster results with no need for reincubation of negatives with Thermo Scientific™ Spectra™ MRSA, a new, improved chromogenic screening plate for Methicillin Resistant Staphylococcus aureus (MRSA). The medium combines excellent sensitivity with the simplicity of a chromogenic plate to create an invaluable tool in the implementation of infection control policies and procedures.
Limit the spread of MRSA by incorporating an effective screening protocol as part of your infection control program. Colonized patients must be targeted for isolation and appropriate treatment as early as possible; therefore, speed and accuracy of results are recognized as critical. Save time and infection control resources by giving negative patients the all-clear faster and more reliably.
  • Rapidly and economically detects nasal colonization of MRSA and MRSA from positive blood cultures
  • Aids in the prevention of MRSA infections
  • Delivers high Positive Predictive Value (PPV) to reduce unnecessary costs
  • Enables quick, efficient screening of patients
  • Produces easy-to-read, denim blue colonies on an opaque background
  • Requires no special handling for optimal workflow 24/7
Spectra MRSA is a selective medium for the screening of clinical samples for the presence of Methicillin-Resistant Staphylococcus aureus (MRSA), with results available within 18 hours and no need to reincubate negatives. The opaque medium incorporates a novel chromogen that yields a blue color as a result of phosphatase activity. This enzyme is present in many staphylococci including Staphylococcus aureus. To allow the differentiation of MRSA accurately, the medium contains a combination of antibacterial compounds designed to inhibit the growth of a wide variety of competitor organisms and MSSA. Also included are compounds to suppress the expression of phosphatase activity in other staphylococci, thus ensuring a high level of sensitivity and specificity.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
ColorOpaque light tan
Fill Volume18 mL
Format85mm Monoplate
Media TypeSpectra MRSA Medium
Packaging TypeFlow Wrapped - Single Layer
Quantity100/Pk.
Manufacturing OriginUnited States
Product TypeAgar
Unit SizeEach
Contents & Storage
2°C to 8°C