Starch Agar w/BCP
Thermo Scientific™
Starch Agar w/BCP
Differentiate Gardnerella vaginalis from other organisms found in the genitourinary tract using Starch Agar w/Brom Cresol Purple.
Have Questions?
Catalog NumberFormat
R0185012mm x 85mm Monoplate
Catalog number R01850
Price (USD)
-
Format:
12mm x 85mm Monoplate
Differentiate Gardnerella vaginalis from other organisms found in the genitourinary tract with Thermo Scientific™ Remel™ Starch Agar w/Brom Cresol Purple. Starch Agar w/Brom Cresol Purple is a modification of peptone starch dextrose (PSD) agar developed by Smith in 19751. Dextrose is replaced with starch and brom cresol purple is added as an indicator to detect hydrolysis of starch by G. vaginalis.
Gardner and Dukes first isolated G. vaginalis from women with nonspecific vaginitis in 19552. When 224 genital specimens were tested, G. vaginalis was the only organism that grew on Starch Agar w/Brom Cresol Purple and fermented starch, with the exceptions of some strains like streptococci and lactobacilli3. The acid produced by the fermentation of starch causes the pH indicator, brom cresol purple, to change the color of the medium from purple to yellow in the zone surrounding growth.
  • Ready-to-use – Convenience of prepared media.
  • Easy to interpret –Yellow color development in the area of growth indicates a positive result. Growth with no color change indicates a negative result.
Starch Agar w/Brom Cresol Purple contains peptone and beef extract that supply nutrients necessary for bacterial growth. Sodium chloride maintains osmotic equilibrium. GCHI Enrichment is a defined supplement which provides vitamins, amino acids, coenzymes and other factors that stimulate growth. Cornstarch neutralizes fatty acids that may be toxic to G. vaginalis and acts as the fermentable substrate in this medium. Brom Cresol Purple is the pH indicator.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
  1. Smith, R.F. 1975. Health Lab Science. 12:219-224.
  2. Gardner, H.L. and C.D. Dukes. 1955. Am. J. Obstet. Gynecol. 69:962-976.
  3. Smith, R.F., J.L. Voss, and E.K. Bailey. 1977. J. Clin. Microbiol. 5:375.
Specifications
DescriptionStarch Agar w/Brom Cresol Purple
Format12mm x 85mm Monoplate
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach