dNTP Set, 100 mM Solutions, 4 x 5 mL - FAQs
dNTP Set, 100 mM Solutions, 4 x 5 mL - FAQs
View additional product information for dNTP Set, 100 mM Solutions - FAQs (R0181, R0182, R0186)
1 product FAQs found
What buffer are the dNTP Set 100 mM Solutions stored in?
The buffer dNTP Set 100 mM Solutions are stored in is water (brought to the correct pH with NaOH).
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.